ENA371177685 Enamine

2,2-dimethyl-2,3-dihydro-1-benzofuran-5-amine

  • MDL number MFCD16708763
  • Cas Number : 31010-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-68
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106892
Availability details for
Quantity
  • Availability
  • Add to Cart