ENA371177688 Enamine

1-{3-[4-(propan-2-yl)phenyl]-1,2,4-oxadiazol-5-yl}ethan-1-amine

  • MDL number MFCD09034290
  • Cas Number : 937666-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106895

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106895
Availability details for
Quantity
  • Availability
  • Add to Cart