ENA371177697 Enamine

3-[(4-fluorobenzene)sulfonamido]naphthalene-2-carboxylic acid

  • MDL number MFCD09043303
  • Cas Number : 926200-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106906

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106906
Availability details for
Quantity
  • Availability
  • Add to Cart