ENA371177706 Enamine

2-[3-(dimethylamino)phenoxy]-5-methylaniline

  • MDL number MFCD08686824
  • Cas Number : 946728-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106978

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106978
Availability details for
Quantity
  • Availability
  • Add to Cart