ENA371177722 Enamine

1-(2,6-dichlorophenyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD11036987
  • Cas Number : 1039931-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216...218
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106999

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106999
Availability details for
Quantity
  • Availability
  • Add to Cart