ENA371177733 Enamine

2-[1-(hydroxymethyl)cyclopropyl]acetic acid

  • MDL number MFCD17011836
  • Cas Number : 869066-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107048

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-107048
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart