ENA371177742 Enamine

3-fluoro-4-(methanesulfonylmethyl)aniline

  • MDL number MFCD20411483
  • Cas Number : 1341641-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107059
Availability details for
Quantity
  • Availability
  • Add to Cart