ENA371177744 Enamine

1-(2-fluoro-4,5-dimethoxyphenyl)ethan-1-one

  • MDL number MFCD17226285
  • Cas Number : 142265-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107061
Availability details for
Quantity
  • Availability
  • Add to Cart