ENA371177745 Enamine

ethyl 5-amino-1-(2-cyanoethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD03618496
  • Cas Number : 1007513-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107062

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107062
Availability details for
Quantity
  • Availability
  • Add to Cart