ENA371177750 Enamine

3-fluoro-5-methoxyaniline

  • MDL number MFCD00077535
  • Cas Number : 2339-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107071
Availability details for
Quantity
  • Availability
  • Add to Cart