ENA371177751 Enamine

2-(3-methyl-1H-pyrazol-1-yl)aniline

  • MDL number MFCD08699295
  • Cas Number : 1006467-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107072

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107072
Availability details for
Quantity
  • Availability
  • Add to Cart