ENA371177757 Enamine

1-[4-(cyclopropylmethoxy)-3-fluorophenyl]ethan-1-amine

  • MDL number MFCD11137192
  • Cas Number : 1019606-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107080
Availability details for
Quantity
  • Availability
  • Add to Cart