ENA371177760 Enamine

1-(4-methanesulfonyl-3-methylphenyl)methanamine

  • MDL number MFCD20721171
  • Cas Number : 694481-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107084
Availability details for
Quantity
  • Availability
  • Add to Cart