ENA371177765 Enamine

5-chloro-3-methyl-1-benzofuran

  • MDL number MFCD06797625
  • Cas Number : 1125-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100269

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100269
Availability details for
Quantity
  • Availability
  • Add to Cart