ENA371177768 Enamine

2-chloro-1,3-benzothiazole-6-sulfonyl chloride

  • MDL number MFCD10575003
  • Cas Number : 6608-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107130

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-107130
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart