ENA371177776 Enamine

1-(2-methylpropyl)pyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD22375328
  • Cas Number : 1384805-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107145
Availability details for
Quantity
  • Availability
  • Add to Cart