ENA371177780 Enamine

3-ethyl-3-methylpyrrolidine

  • MDL number MFCD16737766
  • Cas Number : 34971-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105323
Availability details for
Quantity
  • Availability
  • Add to Cart