ENA371177781 Enamine

3-bromopyrazine-2-carbonitrile

  • MDL number MFCD16842970
  • Cas Number : 1250022-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44...46
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107155

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-107155
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart