ENA371177785 Enamine

1-(2-methylpropyl)cyclopentane-1-carbonitrile

  • MDL number MFCD21227570
  • Cas Number : 1384583-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107161

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107161
Availability details for
Quantity
  • Availability
  • Add to Cart