ENA371177793 Enamine

1lambda6,2,4-thiadiazinane-1,1-dione

  • MDL number MFCD00865125
  • Cas Number : 38668-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107175
Availability details for
Quantity
  • Availability
  • Add to Cart