ENA371177796 Enamine

1-acetyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid

  • MDL number MFCD11180946
  • Cas Number : 51552-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-107178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107178
Availability details for
Quantity
  • Availability
  • Add to Cart