ENA371177799 Enamine

cyclooctanecarbonitrile

  • MDL number MFCD11641481
  • Cas Number : 40636-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107240
Availability details for
Quantity
  • Availability
  • Add to Cart