ENA371177814 Enamine

ethyl({1-[4-(3-methoxyphenyl)phenyl]ethyl})amine

  • MDL number MFCD12860733
  • Cas Number : 1182736-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107265
Availability details for
Quantity
  • Availability
  • Add to Cart