ENA371177815 Enamine

(4-bromophenyl)(cyclobutyl)methanol

  • MDL number MFCD12786812
  • Cas Number : 1216219-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107266

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107266
Availability details for
Quantity
  • Availability
  • Add to Cart