ENA371177818 Enamine

methyl({1-[4'-(propan-2-yloxy)-[1,1'-biphenyl]-3-yl]ethyl})amine

  • MDL number MFCD22375337
  • Cas Number : 1178402-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107269

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107269
Availability details for
Quantity
  • Availability
  • Add to Cart