ENA371177828 Enamine

3-(4-fluorophenyl)azetidine hydrochloride

  • MDL number MFCD08460712
  • Cas Number : 1203681-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-127
Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107291

Safety Information

pcodes P273,P501
MSDS https://www.enaminestore.com/catalog/EN300-107291
hstatements H412
Availability details for
Quantity
  • Availability
  • Add to Cart