ENA371177838 Enamine

2-oxo-1,2-dihydroquinoline-4-carbonitrile

  • MDL number MFCD18803540
  • Cas Number : 63158-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-107372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107372
Availability details for
Quantity
  • Availability
  • Add to Cart