ENA371177852 Enamine

2-hydroxybenzene-1-carboximidamide hydrochloride

  • MDL number MFCD09910296
  • Cas Number : 501904-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107386

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107386
Availability details for
Quantity
  • Availability
  • Add to Cart