ENA371177856 Enamine

1-cyclopropyl-5-oxopyrrolidine-3-carbonitrile

  • MDL number MFCD10004075
  • Cas Number : 1017457-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107392

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107392
Availability details for
Quantity
  • Availability
  • Add to Cart