ENA371177860 Enamine

1-chloro-4-methanesulfonylbutane

  • MDL number MFCD11934617
  • Cas Number : 53394-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107398
Availability details for
Quantity
  • Availability
  • Add to Cart