ENA371177870 Enamine

3-(4-ethylphenyl)-1,2,4-oxadiazole-5-thiol

  • MDL number MFCD17259830
  • Cas Number : 1339756-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107458
Availability details for
Quantity
  • Availability
  • Add to Cart