ENA371177872 Enamine

5-fluoro-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD20441929
  • Cas Number : 1221502-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172...174
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107462

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107462
Availability details for
Quantity
  • Availability
  • Add to Cart