ENA371177878 Enamine

1-(1-phenylethyl)-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD11202268
  • Cas Number : 63261-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107490

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107490
Availability details for
Quantity
  • Availability
  • Add to Cart