ENA371177884 Enamine

4,6-dichloro-2-(3-chloro-4-fluorophenyl)pyrimidine

  • MDL number MFCD18599788
  • Cas Number : 83217-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105549
Availability details for
Quantity
  • Availability
  • Add to Cart