ENA371177892 Enamine

3-(3-methylbutyl)piperidine-2,6-dione

  • MDL number MFCD19584156
  • Cas Number : 1384677-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107514
Availability details for
Quantity
  • Availability
  • Add to Cart