ENA371177897 Enamine

methyl 4-chloro-3-fluoropyridine-2-carboxylate

  • MDL number MFCD16293406
  • Cas Number : 1034921-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107519
Availability details for
Quantity
  • Availability
  • Add to Cart