ENA371177900 Enamine

2-{hydroxy[(pyrazin-2-yl)amino]methylidene}-2,3-dihydropyridin-3-one

  • Cas Number : 1955564-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107522
Availability details for
Quantity
  • Availability
  • Add to Cart