ENA371177903 Enamine

4-[(oxiran-2-yl)methoxy]-1H-indole

  • MDL number MFCD09834410
  • Cas Number : 35308-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-67
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107525
Availability details for
Quantity
  • Availability
  • Add to Cart