ENA371177908 Enamine

4-bromo-3-(4-methylphenyl)-1H-pyrazole

  • MDL number MFCD11855866
  • Cas Number : 1116093-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107532

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107532
Availability details for
Quantity
  • Availability
  • Add to Cart