ENA371177909 Enamine

methyl 2-(methylamino)-2-(3,4,5-trifluorophenyl)acetate

  • MDL number MFCD17284725
  • Cas Number : 1310095-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107533
Availability details for
Quantity
  • Availability
  • Add to Cart