ENA371177911 Enamine

4-methoxy-2,3-dihydro-1H-inden-1-amine

  • MDL number MFCD07373957
  • Cas Number : 52372-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-42
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107546
Availability details for
Quantity
  • Availability
  • Add to Cart