ENA371177918 Enamine

methyl 8-aminooctanoate hydrochloride

  • MDL number MFCD22375357
  • Cas Number : 77420-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107554

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107554
Availability details for
Quantity
  • Availability
  • Add to Cart