ENA371177921 Enamine

1-(4-chlorophenyl)-3-methoxypropan-1-amine hydrochloride

  • MDL number MFCD22375359
  • Cas Number : 1384744-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107557

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107557
Availability details for
Quantity
  • Availability
  • Add to Cart