ENA371177930 Enamine

1-(5-phenyl-1,2,4-oxadiazol-3-yl)ethan-1-amine

  • MDL number MFCD14631142
  • Cas Number : 1292180-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107668

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107668
Availability details for
Quantity
  • Availability
  • Add to Cart