ENA371177939 Enamine

1-(3-bromophenyl)-4-ethyl-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD14587119
  • Cas Number : 1292926-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107681
Availability details for
Quantity
  • Availability
  • Add to Cart