ENA371177951 Enamine

5-bromofuran-3-carboxamide

  • MDL number MFCD20542895
  • Cas Number : 189330-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165...167
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107723

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107723
Availability details for
Quantity
  • Availability
  • Add to Cart