ENA371177953 Enamine

5-ethynyl-2,3-dihydro-1H-indene

  • MDL number MFCD17281926
  • Cas Number : 132464-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107725
Availability details for
Quantity
  • Availability
  • Add to Cart