ENA371177954 Enamine

2-(cycloheptyloxy)propanoic acid

  • MDL number MFCD12784905
  • Cas Number : 1248229-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107726
Availability details for
Quantity
  • Availability
  • Add to Cart