ENA371177956 Enamine

1-amino-2-(4-bromophenyl)butan-2-ol

  • MDL number MFCD11920412
  • Cas Number : 1181734-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107728
Availability details for
Quantity
  • Availability
  • Add to Cart