ENA371177958 Enamine

(2S)-1-[(propan-2-yl)carbamoyl]pyrrolidine-2-carboxylic acid

  • MDL number MFCD10011777
  • Cas Number : 73096-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107731
Availability details for
Quantity
  • Availability
  • Add to Cart